Specifications:- LC Application Compatibility: Bio-compatible. LC Module Type Compatibility: Pump. LC System Compatibility: 1290 inf inity II Bio LC System. LC Use With Instrument: G7132A. Mixer Material: MP35N. Volume: 380 uL. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- LC Application Compatibility: Bio-compatible. LC Module Type Compatibility: Pump. LC System Compatibility: 1290 inf inity II Bio LC System. LC Use With Instrument: G7132A. Mixer Material: MP35N. Volume: 380 uL. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- LC Application Compatibility: Bio-compatible. LC Module Type Compatibility: Pump. LC System Compatibility: 1290 inf inity II Bio LC System. LC Use With Instrument: G7132A. Mixer Material: MP35N. Volume: 380 uL. 1 * 1 Pce HEWLG7132-68380 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou