Specifications:- LC Application Compatibility: Preparative. LC Module Type Compatibility: Pump. LC System Compatibility: 1290 Infini ty II Purification system. LC Use With Instrument: G7161B. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- LC Application Compatibility: Preparative. LC Module Type Compatibility: Pump. LC System Compatibility: 1290 Infini ty II Purification system. LC Use With Instrument: G7161B. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- LC Application Compatibility: Preparative. LC Module Type Compatibility: Pump. LC System Compatibility: 1290 Infini ty II Purification system. LC Use With Instrument: G7161B. 1 * 1 Pce HEWLG7161-68740 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou