Specifications:- Chemical Compatibility: Aqueous Aromatic organics Ketone solvents. Notes: Twister style (double pass) glass cycloni c with thermal conductive polymer encapsulation for use with IsoMist for MP-AES (LoL configuration) Will not fit the 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Chemical Compatibility: Aqueous Aromatic organics Ketone solvents. Notes: Twister style (double pass) glass cycloni c with thermal conductive polymer encapsulation for use with IsoMist for MP-AES (LoL configuration) Will not fit the 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Chemical Compatibility: Aqueous Aromatic organics Ketone solvents. Notes: Twister style (double pass) glass cycloni c with thermal conductive polymer encapsulation for use with IsoMist for MP-AES (LoL configuration) Will not fit the 1 * 1 Pce HEWLG8007-68001 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou