Torches for 5100 SVDV/VDV/RV
Fournisseur: Agilent
G8010-60237 G8012-60000 G8010-60236 G8010-60228 G8010-60231 G8010-60232 G8010-60227 G8010-60234 G8010-60263 G8010-60264 G8010-67000 G8010-68021
HEWLG8010-60237EA 952 EUR InStock
HEWLG8010-60237 HEWLG8012-60000 HEWLG8010-60236 HEWLG8010-60228 HEWLG8010-60231 HEWLG8010-60232 HEWLG8010-60227 HEWLG8010-60234 HEWLG8010-60263 HEWLG8010-60264 HEWLG8010-67000 HEWLG8010-68021
Torches for 5100 SVDV/VDV/RV
Matériel de chromatographie Chromatographie, Pièces de rechange
The 5100 ICP-OES features a plug and play torch, and a simple and effective torch-loader mechanism. This automatically aligns the torch and connects the gases to ensure fast start-up and reproducible performance.
  • Torch allows the 5100 ICP-OES to achieve detection limits of an axially viewed plasma
  • Handles even the toughest samples and achieves unmatched long term stability
Once loaded, there is no need for further adjustments of the torch, or optical alignment of the axial viewing position. Automatic alignment ensures reproducible performance is achieved with each operator, and helps to greatly reduce instrument to instrument variability. The 5100 ICP-OES also features a robust vertical torch in all three available configurations of the instrument. The vertical torch orientation allows you to measure the most challenging samples - from high matrix to volatile organic solvents. This means uncompromised, robust measurements on tough samples with less cleaning, and less downtime. With the longer lifetime expected from vertical torches, less replacement torches are needed.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité