Specifications:- Chemical Compatibility: Aqueous HF acid. Injector Inner Diameter (ID): 1.8 mm. Spectroscopy Instrument Compatibilit y: 5100 5110 5800 5900. Spray Chamber Connection: Ball joint. Torch Demountability: Fully demountable. Torch Injector 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Chemical Compatibility: Aqueous HF acid. Injector Inner Diameter (ID): 1.8 mm. Spectroscopy Instrument Compatibilit y: 5100 5110 5800 5900. Spray Chamber Connection: Ball joint. Torch Demountability: Fully demountable. Torch Injector 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Chemical Compatibility: Aqueous HF acid. Injector Inner Diameter (ID): 1.8 mm. Spectroscopy Instrument Compatibilit y: 5100 5110 5800 5900. Spray Chamber Connection: Ball joint. Torch Demountability: Fully demountable. Torch Injector 1 * 1 Pce HEWLG8020-68003 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou