Specifications:- Chemical Compatibility: Aqueous Organic solvents. Inner Diameter (ID): 1 mm. Length: 1400 mm. Spectroscopy Instrume nt Compatibility: 5100 5110 7700 7800 7900 8800 5800 5900 8900 4100 4200 4210 700 series Liberty Vista series 7850. S 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Chemical Compatibility: Aqueous Organic solvents. Inner Diameter (ID): 1 mm. Length: 1400 mm. Spectroscopy Instrume nt Compatibility: 5100 5110 7700 7800 7900 8800 5800 5900 8900 4100 4200 4210 700 series Liberty Vista series 7850. S 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Chemical Compatibility: Aqueous Organic solvents. Inner Diameter (ID): 1 mm. Length: 1400 mm. Spectroscopy Instrume nt Compatibility: 5100 5110 7700 7800 7900 8800 5800 5900 8900 4100 4200 4210 700 series Liberty Vista series 7850. S 1 * 1 Pce HEWLG8494-60007 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou