Specifications:- LC Application Compatibility: Preparative LC. LC Module Type Compatibility: Fraction Collector. LC Supplies Brand: InfinityLab. LC System Compatibility: 1290 Infinity II Purification system. LC Use With Instrument: G7158B G7159B. Tec 1 * 1 Pce
Fournisseur: Agilent
Specifications:- LC Application Compatibility: Preparative LC. LC Module Type Compatibility: Fraction Collector. LC Supplies Brand: InfinityLab. LC System Compatibility: 1290 Infinity II Purification system. LC Use With Instrument: G7158B G7159B. Tec 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- LC Application Compatibility: Preparative LC. LC Module Type Compatibility: Fraction Collector. LC Supplies Brand: InfinityLab. LC System Compatibility: 1290 Infinity II Purification system. LC Use With Instrument: G7158B G7159B. Tec 1 * 1 Pce HEWLG9321-60051 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou