Specifications:- Carbon Load: 22 %. Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 21.2 mm. LC Colum n Brand: Pursuit XRs. LC Column Maximum Temperature: 60 C . LC Column Phase: C18. LC Platform: Preparative HPLC. Leng 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Carbon Load: 22 %. Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 21.2 mm. LC Colum n Brand: Pursuit XRs. LC Column Maximum Temperature: 60 C . LC Column Phase: C18. LC Platform: Preparative HPLC. Leng 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Carbon Load: 22 %. Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 21.2 mm. LC Colum n Brand: Pursuit XRs. LC Column Maximum Temperature: 60 C . LC Column Phase: C18. LC Platform: Preparative HPLC. Leng 1 * 1 Pce HEWLINF6000150X212 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou