Plgel MIXED-E, 300 x 10 mm, 3 um. Specifications:- Guard Column: No. Inner Diameter (ID): 10 mm. LC Column Brand: PLgel. LC Column M aximum Temperature: 110 C . Length: 300 mm. Molecular Weight Lower Limit: 0 Da. Molecular Weight Upper Limit: 30000 D 1 * 1 Pce
Fournisseur: Agilent
Plgel MIXED-E, 300 x 10 mm, 3 um. Specifications:- Guard Column: No. Inner Diameter (ID): 10 mm. LC Column Brand: PLgel. LC Column M aximum Temperature: 110 C . Length: 300 mm. Molecular Weight Lower Limit: 0 Da. Molecular Weight Upper Limit: 30000 D 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Plgel MIXED-E, 300 x 10 mm, 3 um. Specifications:- Guard Column: No. Inner Diameter (ID): 10 mm. LC Column Brand: PLgel. LC Column M aximum Temperature: 110 C . Length: 300 mm. Molecular Weight Lower Limit: 0 Da. Molecular Weight Upper Limit: 30000 D 1 * 1 Pce HEWLPL1010-6300 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou