PLgel MIXED-LS column
Fournisseur: Agilent
PL1110-1120
HEWLPL1110-1120EA 632 EUR InStock
HEWLPL1110-1120
PLgel MIXED-LS column
Colonnes pour chromatographie
Eliminates particle leakage to improve data quality with light scattering detection. Typical applications: polyethylenes, polyolefins. Compatible with organic solvents.
  • Obtain an instant improvement in data quality
  • No need for conditioning, saving time and solvent costs
  • Maximize the performance of light scattering detectors with minimal investment
The PLgel MIXED-LS series is a PS/DVB packing using an innovative polymerisation technique to virtually eliminate the nano-particle leakage from large pore gels that interferes with light scattering measurement. An immediate improvement in data quality and lower limit of detection is achieved with PLgel MIXED-LS columns in place of conventional GPC columns.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité