PL HFIPgel, 4.6 x 250 mm, 9 um, HPLC column. Specifications:- Guard Column: No. Inner Diameter (ID): 4.6 mm. LC Column Brand: PLgel. LC Column Maximum Temperature: 50 C (HFIP) . Length: 250 mm. Molecular Weight Lower Limit: 200 Da. Molecular Weight 0 1 * 1 Pce
Fournisseur: Agilent
PL HFIPgel, 4.6 x 250 mm, 9 um, HPLC column. Specifications:- Guard Column: No. Inner Diameter (ID): 4.6 mm. LC Column Brand: PLgel. LC Column Maximum Temperature: 50 C (HFIP) . Length: 250 mm. Molecular Weight Lower Limit: 200 Da. Molecular Weight 0 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
PL HFIPgel, 4.6 x 250 mm, 9 um, HPLC column. Specifications:- Guard Column: No. Inner Diameter (ID): 4.6 mm. LC Column Brand: PLgel. LC Column Maximum Temperature: 50 C (HFIP) . Length: 250 mm. Molecular Weight Lower Limit: 200 Da. Molecular Weight 0 1 * 1 Pce HEWLPL1514-5900HFI 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou