Hi-Plex Ca, 4.6 x 250 mm. Specifications:- Endcapped: No. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: Hi-Plex. LC Column Maximum Temperature: 90 C . LC Column Phase: Ca. Length: 250 mm. Molecular Weight Low 1 * 1 Pce
Fournisseur: Agilent
Hi-Plex Ca, 4.6 x 250 mm. Specifications:- Endcapped: No. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: Hi-Plex. LC Column Maximum Temperature: 90 C . LC Column Phase: Ca. Length: 250 mm. Molecular Weight Low 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Hi-Plex Ca, 4.6 x 250 mm. Specifications:- Endcapped: No. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: Hi-Plex. LC Column Maximum Temperature: 90 C . LC Column Phase: Ca. Length: 250 mm. Molecular Weight Low 1 * 1 Pce HEWLPL1570-6810 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou