Specifications:- Guard Column: No. Hardware: SS. Inner Diameter (ID): 4.6 mm. LC Column Brand: AdvanceBio SEC. LC Column Maximum Tem perature: 80 C . LC Column Phase: SEC. Length: 300 mm. Molecular Weight Lower Limit: 1000 Da. Molecular Weight Upper 1 * 3 Pce
Fournisseur: Agilent
Specifications:- Guard Column: No. Hardware: SS. Inner Diameter (ID): 4.6 mm. LC Column Brand: AdvanceBio SEC. LC Column Maximum Tem perature: 80 C . LC Column Phase: SEC. Length: 300 mm. Molecular Weight Lower Limit: 1000 Da. Molecular Weight Upper 1 * 3 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Guard Column: No. Hardware: SS. Inner Diameter (ID): 4.6 mm. LC Column Brand: AdvanceBio SEC. LC Column Maximum Tem perature: 80 C . LC Column Phase: SEC. Length: 300 mm. Molecular Weight Lower Limit: 1000 Da. Molecular Weight Upper 1 * 3 Pce HEWLPL1580-5250K 1 * 3 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou