Specifications:- Guard Column: No. Hardware: PEEK Lined SS. Inner Diameter (ID): 2.1 mm. LC Column Brand: PLRP-S. LC Column Maximum Temperature: 80 C . LC Column Phase: PLRP-S. Length: 50 mm. Particle Size: 5 um. Particle Type: Fully Porous. pH Range 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Guard Column: No. Hardware: PEEK Lined SS. Inner Diameter (ID): 2.1 mm. LC Column Brand: PLRP-S. LC Column Maximum Temperature: 80 C . LC Column Phase: PLRP-S. Length: 50 mm. Particle Size: 5 um. Particle Type: Fully Porous. pH Range 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Guard Column: No. Hardware: PEEK Lined SS. Inner Diameter (ID): 2.1 mm. LC Column Brand: PLRP-S. LC Column Maximum Temperature: 80 C . LC Column Phase: PLRP-S. Length: 50 mm. Particle Size: 5 um. Particle Type: Fully Porous. pH Range 1 * 1 Pce HEWLPL1912-1502PK 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou