Specifications:- Guard Column: No. Hardware: PEEK Lined SS. Inner Diameter (ID): 2.1 mm. LC Column Brand: AdvanceBio SEC. LC Column Maximum Temperature: 80 C (20 to 40 C recommended) . LC Column Phase: SEC. Length: 50 mm. Molecular Weight Lower Limit 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Guard Column: No. Hardware: PEEK Lined SS. Inner Diameter (ID): 2.1 mm. LC Column Brand: AdvanceBio SEC. LC Column Maximum Temperature: 80 C (20 to 40 C recommended) . LC Column Phase: SEC. Length: 50 mm. Molecular Weight Lower Limit 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Guard Column: No. Hardware: PEEK Lined SS. Inner Diameter (ID): 2.1 mm. LC Column Brand: AdvanceBio SEC. LC Column Maximum Temperature: 80 C (20 to 40 C recommended) . LC Column Phase: SEC. Length: 50 mm. Molecular Weight Lower Limit 1 * 1 Pce HEWLPL1980-1250PK 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou