VWR® Hichrom C8 HPLC Columns
Fournisseur: Avantor
HI-3.5C8-100AEA 646 EUR InStock
HI-3.5C8-100A HI-3.5C8-150A HI-3.5C8-50A HI-5C8-100P HICHHI-5C8-10CP HI-5C8-150A HI-5C8-150AF HI-5C8-150AM HI-5C8-150P HI-5C8-250A HI-5C8-250AF HI-5C8-250P HI-5C8-50AS HI-5C8-50P HI-5C8-50SP1 HI-5C8-75P HI-3.5C8-100AF HI-3.5C8-10C5 HI-5C8-100P3 HI-5C8-10C5 HI-5C8-10CP3 HI-5C8-150P3 HI-5C8-150SP1 HI-5C8-250P3 HI-5C8-50P3 HI-5C8-150AE HI-3.5C8-75A
VWR® Hichrom C8 HPLC Columns
Colonnes pour chromatographie
These Avantor® Hichrom C8 columns offer high performance in order to tackle the most challenging reversed-phase applications. The use of ultra pure silica, advanced bonding technology, superior column specification and comprehensive batch validation all contribute to the columns’ excellent reproducibility.
  • Ultra pure, base deactivated porous silica
  • Suitable for acidic, basic and neutral molecules
  • Particle sizes 3,5 and 5 μm available
Combining ultra pure silica with advanced bonding technology results in a densely bonded silica that is both highly robust and highly reproducible. Unpredictable interactions between residual silanol sites on the silica surface and the analyte are effectively eliminated. Comprehensive batch validation ensures absolute batch to batch reproducibility is maintained with acidic, basic and neutral molecules.
Guard cartridges are available for C8 column dimensions, to further extend column lifetime by preventing irreversible adsorption and frit blockage at the top of the column.
Informations de commande: Other dimensions columns and guard cartridges are available on request.
Information de livraison: Supplied with a test chromatogram and batch validation certificate.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité