Avantor® Prevail C8, HPLC Columns
Fournisseur: Avantor
HICH99229EA 1260 EUR InStock
HICH99229
Avantor® Prevail C8, HPLC Columns
Colonnes pour chromatographie
Prevail™ HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases. All phases, apart from Prevail™ Carbohydrate ES are silica-based. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes.
  • Stable from highly organic to highly aqueous
  • Speciality phases for specific applications
  • Excellent sensitivity with microbore and ELSD applications
These columns with stable C8 phase are used for highly hydrophobic compounds that retain too strongly on C18.
Informations sur les accessoires: Use guard cartridges with holder (80101/N) and column coupler (HI-081).
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité