Avantor® Prevail C18, HPLC Columns
Fournisseur: Avantor
HICH43818EA 820 EUR InStock
HICH43818 HICH43871 HICH99200 HICH99322 HICH43829 HICH99202 HICH99204 HICH99279 HICH43903 HICH99208 HICH99209 HICH99210 HICH99211 96683/N 99286/N
Avantor® Prevail C18, HPLC Columns
Colonnes pour chromatographie
Prevail™ HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases. All phases, apart from Prevail™ Carbohydrate ES are silica-based. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes.
  • Stable from highly organic to highly aqueous
  • Speciality phases for specific applications
  • Excellent sensitivity with microbore and ELSD applications
Flexibility to switch between varied mobile phase conditions to suit a variety of applications. Excellent sensitivity for microbore applications.
Informations sur les accessoires: Use guard cartridges with holder (80101/N) and column coupler (HI-081).
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité