McCartney Glass Bottles, Wide Mouth, With Aluminium Screw Cap
Fournisseur: HIMEDIA
GW162-1X100NO
HIMEGW162-1X100NOEA 66.7 EUR InStock
HIMEGW162-1X100NO
McCartney Glass Bottles, Wide Mouth, With Aluminium Screw Cap
Flacons Flacons en verre
McCartney bottles are specially used to carry culture medium and bacterial growth can be done in these bottles.
  • Autoclavable
  • Made of neutral glass
  • Leak-proof clear glass bottle
  • With aluminium screw cap with rubber liner (grey bromo butyl rubber)
  • Size: 84×31 mm
These bottles along with the cap and rubber liner can be autoclaved easily. Also, these bottles can be sterilised with the medium present inside it.
Application: Chemical laboratory, biological laboratory, microbiological laboratory, diagnosis centres, industrial laboratory and various other laboratories.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité