VWR® Hitachi ChromAssist, data station
Fournisseur: VWR Collection
906-0123EA 1990 EUR InStock
906-0123 906-0124 HITA903-0374 HITA903-0375
VWR® Hitachi ChromAssist, data station
Logiciels Chromatography Software
The newly developed ChromAssist Data Station combines the proven intuitive structure of the Hitachi software family with an innovative new design. As a chromatography data system (CDS), ChromAssist contains all the necessary functions for controlling VWR® Hitachi HPLC and UHPLC instruments, including method creation, sample table, data integration and reporting.
  • New user-friendly design
  • Integrated training videos and help functions
  • Simple creation of methods, sample tables
  • Innovative report editor
  • Advanced GLP functionality
Additionally, someMore Product Information
Commander maintenant
COMMANDE
SPÉCIFICATIONS
PRODUITS COMPLEMENTAIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité