Chromaster UV 5410 and 5420 low dispersion semi-micro flow cell for use with narrow bore columns, less than 3 mm inner diameter and less than 3 m particle size. Contains: HITA892-0517,,,,CHROMASTER SEMIMICRO CELL UV 5410&5420 HITA893-4451,,,,IN TUBE 1 * 1 Pce
Fournisseur: VWR Collection
Chromaster UV 5410 and 5420 low dispersion semi-micro flow cell for use with narrow bore columns, less than 3 mm inner diameter and less than 3 m particle size. Contains: HITA892-0517,,,,CHROMASTER SEMIMICRO CELL UV 5410&5420 HITA893-4451,,,,IN TUBE 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Chromaster UV 5410 and 5420 low dispersion semi-micro flow cell for use with narrow bore columns, less than 3 mm inner diameter and less than 3 µm particle size. Contains: HITA892-0517,,,,CHROMASTER SEMIMICRO CELL UV 5410&5420 HITA893-4451,,,,IN TUBE 1 * 1 Pce HITA903-0581 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou