Chromaster DAD 5430 low dispersion semi-micro flow cell for use with narrow bore columns, less than 3 mm inner diameter and less than 3 m particle size. Contains: HITA892-0527,,,,CHROMASTER SEMIMICRO CELL 5430 DAD HITA893-4901,,,,IN TUBE D-SM ASSY ( 1 * 1 Pce
Fournisseur: VWR Collection
Chromaster DAD 5430 low dispersion semi-micro flow cell for use with narrow bore columns, less than 3 mm inner diameter and less than 3 m particle size. Contains: HITA892-0527,,,,CHROMASTER SEMIMICRO CELL 5430 DAD HITA893-4901,,,,IN TUBE D-SM ASSY ( 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Chromaster DAD 5430 low dispersion semi-micro flow cell for use with narrow bore columns, less than 3 mm inner diameter and less than 3 µm particle size. Contains: HITA892-0527,,,,CHROMASTER SEMIMICRO CELL 5430 DAD HITA893-4901,,,,IN TUBE D-SM ASSY ( 1 * 1 Pce HITA903-0584 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou