Mini submarine horizontal electrophoresis unit, HE33
Fournisseur: HOEFER ELECTROPHORESIS
HE33-8-1.5
HOEFHE33-8-1.5EA 453 EUR InStock
HOEFHE33-8-1.5
Mini submarine horizontal electrophoresis unit, HE33
Systèmes d'électrophorèse Cuves d'électrophorèse horizontale
The HE33 mini horizontal electrophoresis unit is designed for very fast separations of DNA restriction fragments in agarose gels.
  • Run a 7×10 cm gel in as little as 5 minutes
  • UV transparent running tray allows the user to image the gel without risk of damage due to handling
  • Variety of comb sizes enables customisation of sample throughput to fit laboratory requirements
  • Adjustable comb height gives complete control over well depth
To achieve passive cooling, simply fill the clear moulded base with 50% ethylene glycol coolant solution, seal it, and place it in a freezer or ice bath until ready to use.
Informations sur les accessoires: All combs require HE31-BK comb back, ordered separately.
Information de livraison: Complete system includes buffer chamber assembly, safety lid with high voltage leads, running tray, casting tray, 4× foam gaskets, bubble level, 1,5 mm thick 8-well comb, and comb back. Basic unit does not include combs or comb back, which can be ordered separately.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
SERVICES
Services: Besoin d'une installation, d'un IQOQ, d'un entretien ou d'une réparation? Contactez-nous : technical.service.be@vwr.com
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité