51781-6X1L
HONC51781-6X1LEA
1860
EUR
InStock
HONC51781-6X1L
N,N-Diméthylformamide ≥99.9%, CHROMASOLV™ en phase gazeuse Headspace, Riedel-de Haën™
N,N-Diméthylformamide
Honeywell Chromasolv™ GC-headspace solvents:
- Developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
- Very pure solvents with extremely low concentrations of the defined residual solvents