Secondary Drinking Water Metals Certified Reference Material, 500 mg/L Zn, 100 mg/L Cu, 30 mg/L Fe, 5 mg/L Mn in HNO₃ 2%, Fluka™
Fournisseur: Honeywell Chemicals
E2165-100ML
HONCE2165-100MLEA 171 EUR InStock
HONCE2165-100ML
Secondary Drinking Water Metals Certified Reference Material, 500 mg/L Zn, 100 mg/L Cu, 30 mg/L Fe, 5 mg/L Mn in HNO₃ 2%, Fluka™
Étalons Étalons cationiques Standards multi-éléments
Honeywell Fluka™ analytical standards offers a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. Our products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité