RS210-204
HONCRS210-204EA
3780
EUR
InStock
HONCRS210-204
n-Heptane ≥99.75%, PRF B&J Brand, meets requirements of ASTM D2699 and D2700, Burdick & Jackson™
n-Heptane
Primary Reference Fuels
- Octane number, the principal performance characteristic of gasoline, is determined through use of certified PRF fuels and a Cooperative Fuel Research (CFR) engine under controlled test conditions
- used in both research and analytical applications
- thorough quality control testing to ensure each lot conforms to the specifications defined in ASTM D2699 for Motor Octane Number (MON) and ASTM 2700 for Research Octane Number (RON).