Ice Cubers, Self Contained, IM
Fournisseur: HOSHIZAKI
IM-100CNE-HC23 IM-100CNE-HC
HOSHIM-100CNE-HC23EA 4130 EUR InStock
HOSHIM-100CNE-HC23 HOSHIM-100CNE-HC
Ice Cubers, Self Contained, IM
Machines à glace
Self contained cube ice makers producing high-quality cube ice. Meeting even the strictest hygiene requirements, this true allrounder offers many application possibilities in segments including the foodservice and medical sector.
  • Uses refrigerant R290, this natural refrigerant is not only climate-friendly, but also boosts the machine, when it comes to performance and production capacity
  • The exterior is made of stainless steel
  • The closed water circuit provides maximum protection against any type of contamination during the entire cubing process
  • Magnetic
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité