HT901-8FOZ
HT901-8FOZEA
70.7
EUR
InStock
HT901-8FOZ
Violet cristallisé solution, Sigma-Aldrich®
Violet cristallisé
Kit component of Gram Stain kits HT90A and HT90T (for staining of heat-fixed films and tissue, respectively).
HT901 is composed of 2.3% certified crystal violet, 0.1% ammonium oxalate monohydrate, and 20% ethyl alcohol, SD3A.