Standard plates and mats, 96-well, WebSeal™
Fournisseur: Thermo Fisher Scientific
60180-P100 60180-P103 60180-P104 60180-P105 60180-P130 60180-P133 60180-P135
HYPE60180-P100EA 80.8 EUR InStock
HYPE60180-P100 HYPE60180-P103 HYPE60180-P104 HYPE60180-P105 HYPE60180-P130 HYPE60180-P133 HYPE60180-P135
Standard plates and mats, 96-well, WebSeal™
Microplaques Plaques d'extraction pour phase liquide
WebSeal™ Standard Plates are plastic, non-coated, nonsterile, and chromatography tested.
  • WebSeal™ plates are manufactured from a GC/MS tested polypropylene material
  • Microplates are chemically and thermally resistant and will tolerate temperatures from will tolerate temperatures from −80 to 121 °C
  • SBS and ANSI standard footprint design for broad instrument compatibility
  • U- and V-bottom wells for optimal sample recovery
  • Low binding PP for homogenous assays and storage
WebSeal™ Mats and Sealing Tapes are nonsterile. Mats are manufactured of EVA (ethylene-vinyl acetate) or pure silicone with or without PTFE coating. Silicone mats are dry heat autoclavable, offer excellent chemical compatibility, and withstand temperatures as low as −80 °C. They resist coring and tearing and provide superior resealability after multiple injections. Sealing tapes minimize evaporation and protect samples from contamination and spills. A wide range of adhesive seals are available for every assay. The thin, lightweight tapes seal onto the plate with a convenient, handheld applicator.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité