Screw caps, 9 mm, level 3 high performance applications, SureSTART™
Fournisseur: Thermo Fisher Scientific
6PMSC9ST1 6PSC9ST1 6PSC9ST101 6PSC9ST101X 6PSC9ST1F 6PSC9ST1R 6PSC9ST1RF 6PSC9ST1RT 6PSC9ST1T 6PSC9STB1 6PSC9STB1F 6PSC9STB1T 6PSC9STBS1 6PSC9STBS1F 6PSC9STBS1T 6PSC9STS1 6PSC9STS1F 6PSC9STS1R 6PSC9STS1RT 6PSC9STS1T 6PSC9TST 6PSC9TSTF 6PSC9TSTR 6PSC9TSTRF 6PSC9TSTRT 6PSC9TSTT 6PSC9STS1Y 6PSC9STS1G
HYPE6PMSC9ST1EA 67.9 EUR InStock
HYPE6PMSC9ST1 HYPE6PSC9ST1 HYPE6PSC9ST101 HYPE6PSC9ST101X HYPE6PSC9ST1F HYPE6PSC9ST1R HYPE6PSC9ST1RF HYPE6PSC9ST1RT HYPE6PSC9ST1T HYPE6PSC9STB1 HYPE6PSC9STB1F HYPE6PSC9STB1T HYPE6PSC9STBS1 HYPE6PSC9STBS1F HYPE6PSC9STBS1T HYPE6PSC9STS1 HYPE6PSC9STS1F HYPE6PSC9STS1R HYPE6PSC9STS1RT HYPE6PSC9STS1T HYPE6PSC9TST HYPE6PSC9TSTF HYPE6PSC9TSTR HYPE6PSC9TSTRF HYPE6PSC9TSTRT HYPE6PSC9TSTT HYPE6PSC9STS1Y HYPE6PSC9STS1G
Screw caps, 9 mm, level 3 high performance applications, SureSTART™
Capuchons Bouchons pour flacons de chromatographie
Use Thermo Scientific™ SureSTART™ 9 mm screw caps with screw vials that have a 9 mm opening, including SureSTART 2 ml glass screw vials, 2 ml gold-grade glass screw vials, total recovery glass screw microvials for <2 ml samples, 1,7 ml high recovery glass screw vials, and glass screw microvials for <2 ml samples (Level 3). Select Level 3 products for ultimate performance and sensitivity from methods, while maintaining the highest levels of sample security and integrity.
Choose from polypropylene caps with all-purpose silicone/PTFE septa designed to reduce autosampler needle issues. Pre-slit and bonded septa options are available. AVCS™ caps incorporate advanced vial closure system technology to ensure optimal seal compression when closing a vial. Use AVCS screw caps with SureStop™ vials for effective seals, consistent data, and increased sample throughput.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité