PTFE/rubber laminated discs
Fournisseur: Thermo Fisher Scientific
TS-12412 TS-12418 TS-12422
HYPETS-12412EA 91.9 EUR InStock
HYPETS-12412 HYPETS-12418 HYPETS-12422
PTFE/rubber laminated discs
Septa
These discs are designed to provide a highly inert and unreactive seal. Constructed of white pharmaceutical rubber with 0,1 mm of PTFE coating bonded to one side, they are excellent for use as cap liners when a highly inert non reactive seal is desired.
  • Autoclavable with no loss of integrity after heating above 100 °C for five hours
Discs are rigid and more difficult to puncture than PTFE/silicone discs.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité