Small reaction vials, Reacti-Vial™
Fournisseur: Thermo Fisher Scientific
TS-13225 TS-13100 TS-13220 TS-13221 TS-13097 TS-13223 TS-13222 TS-13099
HYPETS-13225EA 441 EUR InStock
HYPETS-13225 HYPETS-13100 HYPETS-13220 HYPETS-13221 HYPETS-13097 HYPETS-13223 HYPETS-13222 HYPETS-13099
Small reaction vials, Reacti-Vial™
Fioles
These small reaction vials have an internal cone designed to make small-sample collection and handling easy and convenient. The cone feature is particularly useful for removing small quantities of sample with a syringe.
  • Extra thick glass wall magnifies the sample ideal for observing chemical reactions
  • Amber vials are light-sensitive to well protect the compounds
These vials are ideal for residue isolation, derivative preparation, maximum sample retrieval, moisture protection, sample storage, purification, precipitations, centrifugations and solvent separations.
Information de livraison: Vials are supplied complete with open-top screw caps and teflon/rubber laminated discs.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité