0210204325
ICNA0210204325EA
266
EUR
InStock
ICNA0210204325
Indole-3-butyric acid potassium salt ≥98%, en poudre
Indole-3-butyric acid potassium salt
Indole-3-butyric acid is auxin-family plant hormone.
IBA is thought to be a precursor of indole-3-acetic acid (IAA) the most abundant and the basic auxin natively occurring and functioning in plants. IAA generates the majority of auxin effects in intact plants, and is the most potent native auxin.
Soluble in water (5% w/v).
Attention: Research Use Only (RUO). Protect from light.