0210272001
ICNA0210272001EA
39.7
EUR
InStock
ICNA0210272001
Pregnenolone acetate, poudre blanc cassé
Pregnenolone acetate
One of a small number of "neurosteroids," steroids synthesized in the brain rather than peripheral sources. Decreased levels of these neurosteroids has been implicated in rodent cognitive decline with age, but studies in humans have been contradictory.
Attention: Research Use Only (RUO).