0215179010
ICNA0215179010EA
35.5
EUR
InStock
ICNA0215179010
Acide orotique, anhydre, en poudre
Acide orotique
Soluble in ammonium hydroxide (clear, colorless solution),Sodium hydroxide (50 mg/ml 1 M NaOH); slightly soluble in water (1,7 mg/ml).
Soluble in ammonium hydroxide (clear, colorless solution),Sodium hydroxide (50 mg/ml 1 M NaOH); slightly soluble in water (1,7 mg/ml).