Arthrobacter luteus Yeast lytic enzyme, MP Biomedicals
Fournisseur: MP Biomedicals
0215352610
ICNA0215352610EA 185.5 EUR InStock
ICNA0215352610
Arthrobacter luteus Yeast lytic enzyme, MP Biomedicals
Enzymes
Yeast lytic enzyme is used to digest the cell wall of active yeast cells. It is prepared from a submerged culture of Arthrobacter luteus which effectively lyses cell walls of viable yeast cell.
Lyophilized enzyme partially purified by affinity chromatography
Hydrolyzes poly-β(1→3)-glucose such as yeast cell wall glucan
The primary enzyme in this preparation responsible for lysis of viable yeast cells is β-1,3-glucan laminaripentaohydrolase. It hydrolyzes glucose polymers at the β-1,3-glucan linkages releasing laminaripentaose as the principal product.
Unit definition: One unit will produce a delta A800 of 0,001 per minute at pH 7,5 and 25 °C, using a suspension of brewers yeast as a substrate in a 3 ml reaction mixture.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité