Synonyms:
Lichenin
0215523101
ICNA0215523101EA
113.9
EUR
InStock
ICNA0215523101
Lichenan (de Cetraria islandica), poudre gris pâle
Lichenan
Lichenan is a glucose polysaccharide. It consists of repeating glucose units linked by β-1,3 and β-1,4 glycosidic bonds.
Licheni is used in the study of biochemical and structural characterization of a β-1,3-1,4-glucanase from Bacillus subtilis 168.
Soluble in boiling water and hydrochoride.