0215594490
ICNA0215594490EA
47.4
EUR
InStock
ICNA0215594490
Nordihydroguaiaretic acid, poudre blanche
Nordihydroguaiaretic acid
Inhibitor of lipoxygenases
Inhibitor of lipoxygenases
Soluble in ethanol (5% w/v), methanol, ether, glycol, PEG; insoluble in water
Attention: Research Use Only (RUO).