0215869390
ICNA0215869390EA
44
EUR
InStock
ICNA0215869390
Xylène (mélange d'isomères), liquide clair et incolore ACS
Xylène (mélange d'isomères)
Xylene is used as a solvent. It is used in the laboratory to make baths with dry ice to cool reaction vessels, and as a solvent to remove synthetic immersion oil from the microscope objective in light microscopy.