0219120810
ICNA0219120810EA
36.2
EUR
InStock
ICNA0219120810
D-(+)-Altrose, poudre blanche
D(+)-Altrose
D-(+)-Altrose is used as a substrate to identify, differentiate and characterise aldose isomerases such as L-fucose isomerase from Caldicellulosiruptor saccharolyticus and d-Arabinose isomerase (d-AI) from Bacillus pallidus (B. pallidus) and Klebsiella pneumoniae.
D-(+)-Altrose is used as a substrate to identify, differentiate and characterise aldose isomerases such as L-fucose isomerase from Caldicellulosiruptor saccharolyticus and d-Arabinose isomerase (d-AI) from Bacillus pallidus (B. pallidus) and Klebsiella pneumoniae.
Soluble in water (50 mg/ml - clear, colourless solution); practically insoluble in alcohol.
Attention: Research Use Only (RUO).