Synonyms:
α-D-Glc-(1→4)-α-D-Glc-(1→2)-β-D-Fru, α-Maltosyl β-fructofuranoside
0219866883
ICNA0219866883EA
2076
EUR
InStock
ICNA0219866883
Erlose
Erlose
Erlose is used in studies on dietary preference and utilization of trisose sugars from aphid honeydew by various insects, such as honey bees and ants.
Erlose may be used as a reference compound in assays that analyse the sugars of foods such as royal jelly and honey.
Soluble in water (50 mg/ml)