AlbumiNZ™ Bovine serum albumin (BSA), MP Biomedicals
Fournisseur: MP Biomedicals
0219989990
ICNA0219989990EA 1703 EUR InStock
ICNA0219989990
AlbumiNZ™ Bovine serum albumin (BSA), MP Biomedicals
Protéines et Peptides
AlbumiNZ™ Protease-reduced BSA is useful for immunoassays where proteases may degrade proteins or where peroxidase or alkaline phospahatase may hydrolyse substrates.
AlbumiNZ™ Protease-reduced BSA is chromatographically purified from New Zealand-sourced bovine plasma in an ISO quality system assuring complete traceability and consistent high quality. There is no detectable enzymatic activity.
Biochem/physiol actions: Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité