Synonyms:
2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid
04800681
ICNA04800681EA
68.6
EUR
InStock
ICNA04800681
Acide citrique, anhydre ≥99.5%, poudre cristalline pour l'électrophorèse
Acide citrique
Soluble in water (50 g/ml).
Citric acid acts as an iron chelator. It can support cholesterol/sterol, ubiquinone and isoprenoid biosynthesis in cell culture applications. In animals, citric acid improves the utilisation of nutritional calcium. It is a useful buffer component.