04808196
ICNA04808196EA
210
EUR
InStock
ICNA04808196
3-[(3-Cholamidopropyl)diméthylammonio]-1-propane sulfate (CHAPS) ≥98%, poudre blanche
3-[(3-Cholamidopropyl)diméthylammonio]-1-propane sulfate (CHAPS)
CHAPS is useful for solubilising membrane proteins. Frequently used prior to electrophoresis since it generally does not denature the proteins.
Soluble in 10% aqueous solution (Lit.).
Attention: Research Use Only (RUO).