Anti-Fibrinogen Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fournisseur: MP Biomedicals
0855239
ICNA0855239EA 657 EUR InStock
ICNA0855239
Anti-Fibrinogen Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
The protein encoded by FGA gene is the alpha component of fibrinogen, a blood-borne glycoprotein comprised of three pairs of nonidentical polypeptide chains. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. In addition, various cleavage products of fibrinogen and fibrin regulate cell adhesion and spreading, display vasoconstrictor and chemotactic activities, and are mitogens for several cell types. Mutations in this gene lead to several disorders, including dysfibrinogenemia, hypofibrinogenemia, afibrinogenemia and renal amyloidosis. Alternative splicing results in two isoforms which vary in the carboxy-terminus.
The total protein is measured using the Biuret procedure with bovine albumin as the standard. To ensure that the product titer falls within the required range, antibody titer is standardized by microtiter plate ELISA with human fibrinogen. The product is tested for purity and specificity at final concentration by immunoelectrophoresis. The antibody is predominantly goat IgG; no trace of albumin is detected.
Reconstitute product with 2.0 mL of deionized or distilled water. Gentle swirling may be used to speed rehydration. Do not add sodium azide. Avoid vigorous shaking of the reconstituted material.
Dilution: 1:500 - 1:4,000 for Tissue Staining; 1:1,000 - 1:8,000 for Immuno Blotting; 1:2,000 - 1:16,000 for ELISA/FIA
Type: Primary
Antigen: Fibrinogen
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité