Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-C3 Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Fournisseur: MP Biomedicals
0855385
ICNA0855385EA 336 EUR InStock
ICNA0855385
Anti-C3 Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorps
The complement system provides innate defense against microbial infection and is a "complement" to antibody mediated immunity. The complement system consists of thirty-five interacting plasma and membrane associated proteins which contribute to host-defense and initiate and amplify inflammation, even in the preimmune state where specific antibodies and lymphocytes are not available. In addition to the complement components themselves, this system also contains several soluble factors that prevent spontaneous complement activation from occurring in solution, as well as, several regulatory membrane associated proteins that protect host cells from accidental complement attack. Cleavage of the C3 component by either the classical pathway or the alternative pathway releases C3a and C3b. C3a appears to be important in many inflammatory responses while the C3b fragment covalently binds to the cell or bacterial surface and plays a role in opsonisation. Binding of C3b to the C4b component of the C3 convertase, results in C5 convertase (C4b3b2a) formation.

The total protein is measured using the Biuret procedure with bovine albumin as the standard; F/P ratio is calculated using absorbance at 491.5nm. Antibody titer of the IgG fraction is standardized with an in-house control by immunoelectrophoresis. The IgG fraction is tested for purity and specificity at 40 mg/mL using immunoelectrophoresis; No trace of albumin is detected. Antibody activity to serum albumin is not present.

Reconstitute product with 2.0 mL of deionized or distilled water. Gentle swirling may be used to speed rehydration. Avoid vigorous shaking of the reconstituted material.

Suggested working dilution: Anti-complement immunofluorescent (ACIF) assays: 1:15

Type: Primary
Antigen: C3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Guinea pig

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité