Anti-Ig Goat Antibody (FITC (Fluorescein Isothiocyanate))
Fournisseur: MP Biomedicals
0855526
ICNA0855526EA 292 EUR InStock
ICNA0855526
Anti-Ig Goat Antibody (FITC (Fluorescein Isothiocyanate))
Anticorps
This product is fluorescein-5-isothiocyanate (FITC "Isomer I")-conjugated goat affinity purified F(ab')2 fragments to mouse immunoglobulins (IgG,IgA,IgM) and buffer salts.The total protein is measured by absorbance (A)280 nm); F/P ratio is calculated using absorbance at 491.5nm. Antibody titer of the affinity purified F(ab')2fragments is standardized with an in-house control by immunoelectrophoresis. The affinity purified F(ab')2 fragment is tested for purity at 10 mg/mL using immunoelectrophoresis. The product is goat F(ab')2; no trace of albumin, whole molecule IgG or Fc is detected. This product contains sodium azide. To prevent formation of toxic vapors, do not mix with strongly acidic solutions. To prevent formation of potentially explosive metallic azides in metal plumbing, always wash into drain with copious quantities of water.
Type: Secondary
Antigen: Ig
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: F(ab')2
Reactivity: Mouse
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité