Rat IgM Isotype Control
Fournisseur: MP Biomedicals
0855951
ICNA0855951EA 389 EUR InStock
ICNA0855951
Rat IgM Isotype Control
Anticorps
Product is the lyophilized powder of rat IgG and buffer salts.IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections. The total protein is determined using the Biuret procedure, with bovine albumin as the standard. Each vial is overfilled by approximately 10 percent so that a minimum of 20 mg of rat IgG is obtained upon reconstitution.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité