Protein G, MP Biomedicals
Fournisseur: MP Biomedicals
08672651
ICNA08672651EA 371 EUR InStock
ICNA08672651
Protein G, MP Biomedicals
Protéines purification Protein Purification Resins
Protein G is a monomeric protein lacking cysteine residues. Recombinant Protein G is a highly stable surface receptor that is capable of binding the Fc portion of immunoglobulins, especially IgGs from a large number of species.
  • Freely soluble in water and standard buffers
  • Extinction coefficient: E1% 280=~9,5
  • Particle size 31 kDa
Protein G contains two IgG-binding B regions, binds both Fc and Fab fragments of IgG. Does not cross-react with human albumin. It can be used with immunologically active enzyme conjugates, ELISA, dot blot and Western Blot techniques, immunoprecipitation, and affinity purification of polyclonal and monoclonal antibodies. Colloidal gold conjugated protein G is suitable for electron microscopy application.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité