Cell detachment solution, Accutase, MP Biomedicals
Fournisseur: MP Biomedicals
091000449
ICNA091000449EA 58 EUR InStock
ICNA091000449
Cell detachment solution, Accutase, MP Biomedicals
Suppléments et additifs pour culture cellulaire
Accutase solution is a trypsin replacement that contains both proteolytic and collagenolytic enzymes for cell detachment.
Accutase is effective in detaching primary fibroblasts, endothelial cells, neurons, tumour cell lines, and insect cells. It performs well in detaching cells for analysis of cell surface markers, virus growth assay, and flow cytometry as well as bioreactor scale-up.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité